Anti-BPI Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-4292R-A647
BOSSBS-4292R-A647EA 893 EUR
BOSSBS-4292R-A647
Anti-BPI Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
The antimicrobial protein Bactericidal Permeability Increasing protein (BPI) is a 55 kDa protein found in the primary granules of polymorhonuclear leukocytes (PMN). The cytotoxicity action of BPI is limited to gram negative bacteria, reflecting the high affinity of BPI for bacterial LPS. Binding of BPI to live bacteria via LPS causes anti-infective activites: 1) cytotoxicity via sequential damage to bacterial outer and inner lipid membranes, 2) neutralization of gram-negative bacterial LPS, 3) opsonization of bacteria to enhance phagocytosis by neutrophils.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BPI
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human