Anti-BPI Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-4292R-A680
BOSSBS-4292R-A680EA 890 EUR
BOSSBS-4292R-A680
Anti-BPI Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The antimicrobial protein Bactericidal Permeability Increasing protein (BPI) is a 55 kDa protein found in the primary granules of polymorhonuclear leukocytes (PMN). The cytotoxicity action of BPI is limited to gram negative bacteria, reflecting the high affinity of BPI for bacterial LPS. Binding of BPI to live bacteria via LPS causes anti-infective activites: 1) cytotoxicity via sequential damage to bacterial outer and inner lipid membranes, 2) neutralisation of gram-negative bacterial LPS, 3) opsonization of bacteria to enhance phagocytosis by neutrophils.

Type: Primary
Antigen: BPI
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 141-240/487
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR