Anti-BPI Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-4292R-HRP
BOSSBS-4292R-HRPEA 900 EUR
BOSSBS-4292R-HRP
Anti-BPI Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The antimicrobial protein Bactericidal Permeability Increasing protein (BPI) is a 55 kDa protein found in the primary granules of polymorhonuclear leukocytes (PMN). The cytotoxicity action of BPI is limited to gram negative bacteria, reflecting the high affinity of BPI for bacterial LPS. Binding of BPI to live bacteria via LPS causes anti-infective activites: 1) cytotoxicity via sequential damage to bacterial outer and inner lipid membranes, 2) neutralization of gram-negative bacterial LPS, 3) opsonization of bacteria to enhance phagocytosis by neutrophils.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: BPI
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR