Anti-Defensin alpha 5 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-4313R
BOSSBS-4313REA 725 EUR
BOSSBS-4313R
Anti-Defensin alpha 5 Rabbit Polyclonal Antibody
Antikörper
Has antimicrobial activity against Gram-negative and Gram-positive bacteria. Defensins are thought to kill microbes by permeabilizing their plasma membrane. All DEFA5 peptides exert antimicrobial activities, but their potency is affected by peptide processing.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Defensin alpha 5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR