Anti-Streptococcus suis Suilysin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-4537R-A750
BOSSBS-4537R-A750EA 890 EUR
BOSSBS-4537R-A750
Anti-Streptococcus suis Suilysin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Streptococcus suis suilysin belongs to a family of thiol-activated, membrane damaging toxins produced by some gram-positive bacteria. Although suilysin has been implicated as virulence associated, it has yet to be shown conclusively to be a virulence factor.
Type: Primary
Antigen: Streptococcus suis Suilysin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria