Anti-IL2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-4586R-A555
BOSSBS-4586R-A555EA 893 EUR
BOSSBS-4586R-A555
Anti-IL2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Produced by T-cells in response to antigenic or mitogenic stimulation, this protein is required for T-cell proliferation and other activities crucial to regulation of the immune response. Can stimulate B-cells, monocytes, lymphokine-activated killer cells, natural killer cells, and glioma cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat