Anti-IL2 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-4586R-CY5
BOSSBS-4586R-CY5EA 893 EUR
BOSSBS-4586R-CY5
Anti-IL2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Produced by T-cells in response to antigenic or mitogenic stimulation, this protein is required for T-cell proliferation and other activities crucial to regulation of the immune response. Can stimulate B-cells, monocytes, lymphokine-activated killer cells, natural killer cells, and glioma cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat