Anti-MMP9 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-4593R-A750
BOSSBS-4593R-A750EA 899 EUR
BOSSBS-4593R-A750
Anti-MMP9 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
May play an essential role in local proteolysis of the extracellular matrix and in leukocyte migration. Could play a role in bone osteoclastic resorption. Cleaves KiSS1 at a Gly-|-Leu bond. Cleaves type IV and type V collagen into large C-terminal three quarter fragments and shorter N-terminal one quarter fragments. Degrades fibronectin but not laminin or Pz-peptide.

Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-250/707
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Dog; Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR