Anti-PILR beta Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-4670R-A750
BOSSBS-4670R-A750EA 890 EUR
BOSSBS-4670R-A750
Anti-PILR beta Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Control of cell signaling via SHP-1 is thought to occur through a balance between PILRalpha-mediated inhibition and PILRbeta-mediated activation. These paired immunoglobulin-like receptor genes are located in a tandem head-to-tail orientation on chromosome 7. PILRB is the non-ITIM-bearing member of the receptor pair, which has a truncated cytoplasmic tail relative to its ITIM-bearing partner and functions in the activating role. There are three named isoforms produced by alternative splicing.

Type: Primary
Antigen: PILR beta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/224
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR