Anti-PILRB1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-4670R-CY5.5
BOSSBS-4670R-CY5.5EA 900 EUR
BOSSBS-4670R-CY5.5
Anti-PILRB1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Control of cell signaling via SHP-1 is thought to occur through a balance between PILRalpha-mediated inhibition and PILRbeta-mediated activation. These paired immunoglobulin-like receptor genes are located in a tandem head-to-tail orientation on chromosome 7. PILRB is the non-ITIM-bearing member of the receptor pair, which has a truncated cytoplasmic tail relative to its ITIM-bearing partner and functions in the activating role. There are three named isoforms produced by alternative splicing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PILRB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR