Anti-EBNA 3B Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-4698R-CY7
BOSSBS-4698R-CY7EA 883 EUR
BOSSBS-4698R-CY7
Anti-EBNA 3B Rabbit Polyclonal Antibody (Cy7®)
Antikörper
May be involved in transcriptional regulation of both viral and cellular genes by interacting with the host DNA-binding protein RBPJ.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EBNA 3B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR