Anti-SLCO2A1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-4710R-A488
BOSSBS-4710R-A488EA 893 EUR
BOSSBS-4710R-A488
Anti-SLCO2A1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
May mediate the release of newly synthesized prostaglandins from cells, the transepithelial transport of prostaglandins, and the clearance of prostaglandins from the circulation. Transports PGD2, as well as PGE1, PGE2 and PGF2A.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLCO2A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat