Anti-SLCO2A1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-4710R-A680
BOSSBS-4710R-A680EA 890 EUR
BOSSBS-4710R-A680
Anti-SLCO2A1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
May mediate the release of newly synthesized prostaglandins from cells, the transepithelial transport of prostaglandins, and the clearance of prostaglandins from the circulation. Transports PGD2, as well as PGE1, PGE2 and PGF2A.

Type: Primary
Antigen: SLCO2A1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 611-643/643
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR