Anti-SLCO2A1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-4710R-CY7
BOSSBS-4710R-CY7EA 900 EUR
BOSSBS-4710R-CY7
Anti-SLCO2A1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
May mediate the release of newly synthesized prostaglandins from cells, the transepithelial transport of prostaglandins, and the clearance of prostaglandins from the circulation. Transports PGD2, as well as PGE1, PGE2 and PGF2A.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLCO2A1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR