Anti-LLGL2 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-4771R
BOSSBS-4771REA 704 EUR
BOSSBS-4771R
Anti-LLGL2 Rabbit Polyclonal Antibody
Antikörper
The lethal (2) giant larvae protein of Drosophila plays a role in asymmetric cell division, epithelial cell polarity, and cell migration. This human gene encodes a protein similar to lethal (2) giant larvae of Drosophila. In fly, the protein's ability to localize cell fate determinants is regulated by the atypical protein kinase C (aPKC). In human, this protein interacts with aPKC-containing complexes and is cortically localized in mitotic cells. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Jul 2008].
WB(1:100-1000), IHC-P(1More Product Information