Anti-CD8A Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-4790R-CY3
BOSSBS-4790R-CY3EA 901 EUR
BOSSBS-4790R-CY3
Anti-CD8A Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Identifies cytotoxic/suppressor T-cells that interact with MHC class I bearing targets. CD8 is thought to play a role in the process of T-cell mediated killing. CD8 alpha chains binds to class I MHC molecules alpha-3 domains.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CD8A
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen