Anti-NT5E Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: Bioss
BS-4834R-PE
BOSSBS-4834R-PEEA 944 EUR
BOSSBS-4834R-PE
Anti-NT5E Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Hydrolyzes extracellular nucleotides into membrane permeable nucleosides. Exhibits AMP-, NAD-, and NMN-nucleosidase activities.

Recommended Dilutions: Flow Cytometry: 1:20-100

Type: Primary
Antigen: NT5E
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Dog, Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR