Anti-CD8B Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-4914R-A350
BOSSBS-4914R-A350EA 893 EUR
BOSSBS-4914R-A350
Anti-CD8B Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Identifies cytotoxic/suppressor T-cells that interact with MHC class I bearing targets. CD8 is thought to play a role in the process of T-cell mediated killing.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: CD8B
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen