Anti-CD8B Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-4914R-A680
BOSSBS-4914R-A680EA 890 EUR
BOSSBS-4914R-A680
Anti-CD8B Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Identifies cytotoxic/suppressor T-cells that interact with MHC class I bearing targets. CD8 is thought to play a role in the process of T-cell mediated killing.
Type: Primary
Antigen: CD8B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/210
Host: Rabbit
Isotype: IgG
Reactivity: Human