Anti-CD8B Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-4914R-CY7
BOSSBS-4914R-CY7EA 900 EUR
BOSSBS-4914R-CY7
Anti-CD8B Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Identifies cytotoxic/suppressor T-cells that interact with MHC class I bearing targets. CD8 is thought to play a role in the process of T-cell mediated killing.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD8B
Clonality: Polyclonal
Clone: 0
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR