Anti-Osteocalcin Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-4917R-A750
BOSSBS-4917R-A750EA 899 EUR
BOSSBS-4917R-A750
Anti-Osteocalcin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Osteocalcin belongs to the osteocalcin/matrix Gla protein family and constitutes 1 to 2% of the total bone protein. It is a 49 amino acid single chain vitamin K dependent protein, made by osteoblasts, and is a major component of the noncollagenous bone matrix. Post translational modification by a vitamin K dependent carboxylase produces three gamma carboxyglutamic acid residues at positions 17, 21 and 24, giving it a high affinity for calcium. It also binds strongly to apatite.

Type: Primary
Antigen: Osteocalcin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-100/100
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR