Anti-BGLAP Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-4917R-CY7
BOSSBS-4917R-CY7EA 901 EUR
BOSSBS-4917R-CY7
Anti-BGLAP Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Osteocalcin belongs to the osteocalcin/matrix Gla protein family and constitutes 1 to 2% of the total bone protein. It is a 49 amino acid single chain vitamin K dependent protein, made by osteoblasts, and is a major component of the noncollagenous bone matrix. Post translational modification by a vitamin K dependent carboxylase produces three gamma carboxyglutamic acid residues at positions 17, 21 and 24, giving it a high affinity for calcium. It also binds strongly to apatite.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BGLAP
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR