Anti-BGLAP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-4917R-HRP
BOSSBS-4917R-HRPEA 901 EUR
BOSSBS-4917R-HRP
Anti-BGLAP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Osteocalcin belongs to the osteocalcin/matrix Gla protein family and constitutes 1 to 2% of the total bone protein. It is a 49 amino acid single chain vitamin K dependent protein, made by osteoblasts, and is a major component of the noncollagenous bone matrix. Post translational modification by a vitamin K dependent carboxylase produces three gamma carboxyglutamic acid residues at positions 17, 21 and 24, giving it a high affinity for calcium. It also binds strongly to apatite.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: BGLAP
Clonality: Polyclonal
Clone:
Conjugation:More Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol BGLAP
Antigen-Name Bone gamma-carboxyglutamate (gla) protein
Environmentally Preferable
Antigen-Synonyme OCN|OC|BGP
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation HRP (Horseradish Peroxidase)
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 632
Isotyp IgG
ImmunoChemie Yes
Immunogen 60-100/100
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR