Anti-MT3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-4964R-A488
BOSSBS-4964R-A488EA 876 EUR
BOSSBS-4964R-A488
Anti-MT3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Binds heavy metals. Contains three zinc and three copper atoms per polypeptide chain and only a negligible amount of cadmium. Inhibits survival and neurite formation of cortical neurons in vitro.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MT3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human