Anti-human secretory IgA Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-4964R-A680
BOSSBS-4964R-A680EA 874 EUR
BOSSBS-4964R-A680
Anti-human secretory IgA Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Binds heavy metals. Contains three zinc and three copper atoms per polypeptide chain and only a negligible amount of cadmium. Inhibits survival and neurite formation of cortical neurons in vitro.

Type: Primary
Antigen: human secretory IgA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR