Anti-MT3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-4964R-CY5
BOSSBS-4964R-CY5EA 883 EUR
BOSSBS-4964R-CY5
Anti-MT3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Binds heavy metals. Contains three zinc and three copper atoms per polypeptide chain and only a negligible amount of cadmium. Inhibits survival and neurite formation of cortical neurons in vitro.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MT3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR