Anti-MT3 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-4964R-CY7
BOSSBS-4964R-CY7EA 883 EUR
BOSSBS-4964R-CY7
Anti-MT3 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Binds heavy metals. Contains three zinc and three copper atoms per polypeptide chain and only a negligible amount of cadmium. Inhibits survival and neurite formation of cortical neurons in vitro.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MT3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human