Anti-BDNF Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-4989R-CY3
BOSSBS-4989R-CY3EA 909 EUR
BOSSBS-4989R-CY3
Anti-BDNF Rabbit Polyclonal Antibody (Cy3®)
Antikörper
During development, promotes the survival and differentiation of selected neuronal populations of the peripheral and central nervous systems. Participates in axonal growth, pathfinding and in the modulation of dendritic growth and morphology. Major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. The versatility of BDNF is emphasized by its contribution to a range of adaptive neuronal responses including long-term potentiation (LTP), long-term depression (LTD), certain forms of short-term synaptic plasticity, as well as homeostatic regulation ofMore Product Information