Anti-BDNF Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-4989R-CY5.5
BOSSBS-4989R-CY5.5EA 909 EUR
BOSSBS-4989R-CY5.5
Anti-BDNF Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
During development, promotes the survival and differentiation of selected neuronal populations of the peripheral and central nervous systems. Participates in axonal growth, pathfinding and in the modulation of dendritic growth and morphology. Major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. The versatility of BDNF is emphasized by its contribution to a range of adaptive neuronal responses including long-term potentiation (LTP), long-term depression (LTD), certain forms of short-term synaptic plasticity, as well as homeostatic regulation of intrinsic neuronal excitability.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BDNF
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR