Anti-PGLS Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-5007R-CY5
BOSSBS-5007R-CY5EA 900 EUR
BOSSBS-5007R-CY5
Anti-PGLS Rabbit Polyclonal Antibody (Cy5®)
Antikörper
PGLS belongs to the glucosamine/galactosamine-6-phosphate isomerase family, 6-phosphogluconolactonase subfamily. It is implicated in the hydrolysis of 6-phosphogluconolactone to 6-phosphogluconate.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PGLS
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR