Anti-ABCG5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-5013R-A750
BOSSBS-5013R-A750EA 899 EUR
BOSSBS-5013R-A750
Anti-ABCG5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Transporter that appears to play an indispensable role in the selective transport of the dietary cholesterol in and out of the enterocytes and in the selective sterol excretion by the liver into bile.

Type: Primary
Antigen: ABCG5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/651
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR