Anti-ACADVL Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-5018R-CY3
BOSSBS-5018R-CY3EA 900 EUR
BOSSBS-5018R-CY3
Anti-ACADVL Rabbit Polyclonal Antibody (Cy3®)
Antikörper
ACADVL (acyl-Coenzyme A dehydrogenase, very long chain) catalyzes the first step of the mitochondrial fatty acid beta-oxidation pathway. It is specific to esters of long-chain and very long chain fatty acids such as palmitoyl-CoA and stearoyl-CoA. Deficiencies in ACADVL are associated with reduced myocardial fatty acid beta-oxidation and cardiomyopathy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACADVL
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR