Anti-ACAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-5019R-A555
BOSSBS-5019R-A555EA 893 EUR
BOSSBS-5019R-A555
Anti-ACAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
This gene encodes a mitochondrially localized enzyme that catalyzes the reversible formation of acetoacetyl-CoA from two molecules of acetyl-CoA. Defects in this gene are associated with 3-ketothiolase deficiency, an inborn error of isoleucine catabolism characterized by urinary excretion of 2-methyl-3-hydroxybutyric acid, 2-methylacetoacetic acid, tiglylglycine, and butanone. [provided by RefSeq, Feb 2009].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ACAT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat