Anti-ACAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-5019R-A680
BOSSBS-5019R-A680EA 890 EUR
BOSSBS-5019R-A680
Anti-ACAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This gene encodes a mitochondrially localised enzyme that catalyses the reversible formation of acetoacetyl-CoA from two molecules of acetyl-CoA. Defects in this gene are associated with 3-ketothiolase deficiency, an inborn error of isoleucine catabolism characterised by urinary excretion of 2-methyl-3-hydroxybutyric acid, 2-methylacetoacetic acid, tiglylglycine, and butanone.

Type: Primary
Antigen: ACAT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/427
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR