Anti-ACAT1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-5019R-CY3
BOSSBS-5019R-CY3EA 900 EUR
BOSSBS-5019R-CY3
Anti-ACAT1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
This gene encodes a mitochondrially localized enzyme that catalyzes the reversible formation of acetoacetyl-CoA from two molecules of acetyl-CoA. Defects in this gene are associated with 3-ketothiolase deficiency, an inborn error of isoleucine catabolism characterized by urinary excretion of 2-methyl-3-hydroxybutyric acid, 2-methylacetoacetic acid, tiglylglycine, and butanone. [provided by RefSeq, Feb 2009].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACAT1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR