Anti-SOAT2 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-5020R-CY5.5
BOSSBS-5020R-CY5.5EA 900 EUR
BOSSBS-5020R-CY5.5
Anti-SOAT2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Plays a role in lipoprotein assembly and dietary cholesterol absorption. In addition to its acyltransferase activity, it may act as a ligase. May provide cholesteryl esters for lipoprotein secretion from hepatocytes and intestinal mucosa.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SOAT2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat