Anti-ACOX1 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-5021R
BOSSBS-5021REA 725 EUR
BOSSBS-5021R
Anti-ACOX1 Rabbit Polyclonal Antibody
Antikörper
Defects in ACOX1 are the cause of adrenoleukodystrophy pseudoneonatal (Pseudo-NALD); also known as peroxisomal acyl-CoA oxidase deficiency. Pseudo-NALD is a peroxisomal single-enzyme disorder. Clinical features include mental retardation, leukodystrophy, seizures, mild hepatomegaly, hearing deficit. Pseudo-NALD is characterized by increased plasma levels of very-long chain fatty cids, due to decreased or absent peroxisome acyl-CoA oxidase activity. Peroxisomes are intact and functioning.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ACOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat