Anti-AGPAT1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-5023R
BOSSBS-5023REA 725 EUR
BOSSBS-5023R
Anti-AGPAT1 Rabbit Polyclonal Antibody
Antikörper
AGPAT1 converts lysophosphatidic acid (LPA) into phosphatidic acid by incorporating an acyl moiety at the sn-2 position of the glycerol backbone.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: AGPAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR