Anti-AGPAT1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-5023R-CY7
BOSSBS-5023R-CY7EA 900 EUR
BOSSBS-5023R-CY7
Anti-AGPAT1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
AGPAT1 converts lysophosphatidic acid (LPA) into phosphatidic acid by incorporating an acyl moiety at the sn-2 position of the glycerol backbone.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AGPAT1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR