Anti-ACOT8 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-5031R-CY7
BOSSBS-5031R-CY7EA 900 EUR
BOSSBS-5031R-CY7
Anti-ACOT8 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Acyl-CoA thioesterases are a group of enzymes that catalyze the hydrolysis of acyl-CoAs to the free fatty acid and coenzyme A (CoASH), providing the potential to regulate intracellular levels of acyl-CoAs, free fatty acids and CoASH. Acyl-coenzyme A Thioesterase 8, also known as ACOT8 may mediate Nef-induced down-regulation of CD4. It is a major thioesterase in peroxisomes and competes with BAAT (Bile acid CoA: amino acid N-acyltransferase) for bile acid-CoA substrate (such as chenodeoxycholoyl-CoA). It shows a preference for medium-length fatty acyl-CoAs and may be involved in the metabolic regulation of peroxisome proliferation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ACOT8
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat