Anti-AGPAT4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-5033R-A680
BOSSBS-5033R-A680EA 899 EUR
BOSSBS-5033R-A680
Anti-AGPAT4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Converts lysophosphatidic acid (LPA) into phosphatidic acid by incorporating an acyl moiety at the sn-2 position of the glycerol backbone.

Type: Primary
Antigen: AGPAT4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 231-330/378
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR