Anti-COL4A3BP Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-5044R-A350
BOSSBS-5044R-A350EA 893 EUR
BOSSBS-5044R-A350
Anti-COL4A3BP Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Shelters ceramides and diacylglycerol lipids inside its START domain and mediates the intracellular trafficking of ceramides and diacylglycerol lipids in a non-vesicular manner.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COL4A3BP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR