Anti-COL4A3BP Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-5044R-CY5
BOSSBS-5044R-CY5EA 900 EUR
BOSSBS-5044R-CY5
Anti-COL4A3BP Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Shelters ceramides and diacylglycerol lipids inside its START domain and mediates the intracellular trafficking of ceramides and diacylglycerol lipids in a non-vesicular manner.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: COL4A3BP
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen