Anti-DECR1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-5056R-A350
BOSSBS-5056R-A350EA 893 EUR
BOSSBS-5056R-A350
Anti-DECR1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Auxiliary enzyme of beta-oxidation. It participates in the metabolism of unsaturated fatty enoyl-CoA esters having double bonds in both even- and odd-numbered positions. Catalyzes the NADP-dependent reduction of 2,4-dienoyl-CoA to yield trans-3-enoyl-CoA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DECR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR