Anti-DECR1 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-5056R-CY5.5
BOSSBS-5056R-CY5.5EA 900 EUR
BOSSBS-5056R-CY5.5
Anti-DECR1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Auxiliary enzyme of beta-oxidation. It participates in the metabolism of unsaturated fatty enoyl-CoA esters having double bonds in both even- and odd-numbered positions. Catalyzes the NADP-dependent reduction of 2,4-dienoyl-CoA to yield trans-3-enoyl-CoA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DECR1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat