Anti-ECHS1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-5058R-A680
BOSSBS-5058R-A680EA 890 EUR
BOSSBS-5058R-A680
Anti-ECHS1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
ECHS1 (Enoyl Coenzyme A hydratase, short chain, 1) catalyses the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs, in the second step of the mitochondrial fatty acid beta-oxidation pathway.

Type: Primary
Antigen: ECHS1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 111-210/290
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR