Anti-NMRK1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-5058R-CY3
BOSSBS-5058R-CY3EA 900 EUR
BOSSBS-5058R-CY3
Anti-NMRK1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
ECHS1 (Enoyl Coenzyme A hydratase, short chain, 1) catalyzes the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs, in the second step of the mitochondrial fatty acid beta-oxidation pathway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NMRK1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR