Anti-NMRK1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-5058R-CY7
BOSSBS-5058R-CY7EA 900 EUR
BOSSBS-5058R-CY7
Anti-NMRK1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
ECHS1 (Enoyl Coenzyme A hydratase, short chain, 1) catalyzes the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs, in the second step of the mitochondrial fatty acid beta-oxidation pathway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NMRK1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

TECHNISCHE DATEN

Antigen-Symbol NMRK1
Environmentally Preferable
Antigen-Name Nicotinamide Riboside Kinase 1
Antigen-Synonyme NRK1|bA235O14.2|C9orf95
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy7®
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 54981
Isotyp IgG
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR