Anti-TAAR8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-5060R-A488
BOSSBS-5060R-A488EA 893 EUR
BOSSBS-5060R-A488
Anti-TAAR8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
FADS1 is a component of a lipid metabolic pathway that catalyzes biosynthesis of highly unsaturated fatty acids from precursor essential polyunsaturated fatty acids, linoleic acid and alphanlinolenic acid. It catalyzes the desaturation of dihomo gamma linoleic acid and eicosatetraenoic acid to generate arachidonic acid and eicosapentaenoic acid respectively.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TAAR8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR