Anti-TAAR8 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-5060R-CY5.5
BOSSBS-5060R-CY5.5EA 900 EUR
BOSSBS-5060R-CY5.5
Anti-TAAR8 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
FADS1 is a component of a lipid metabolic pathway that catalyzes biosynthesis of highly unsaturated fatty acids from precursor essential polyunsaturated fatty acids, linoleic acid and alphanlinolenic acid. It catalyzes the desaturation of dihomo gamma linoleic acid and eicosatetraenoic acid to generate arachidonic acid and eicosapentaenoic acid respectively.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TAAR8
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat